Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQD13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339994.1 | NP_001326850.1 | 4 - 5 | 1698 | 83 | |
NM_115831.3 | NP_191528.1 | 5 - 6 | 1217 | 83 | ||
GenBank mRNA | AY128329.1 | - | 5 - 6 | 1020 | 83 | |
BT001176.1 | - | 3 - 4 | 762 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |