Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035819.1 | NP_001030896.1 | 11 - 12 | 4620 | 91 | |
NM_001203209.1 | NP_001190138.1 | 12 - 13 | 4547 | 91 | ||
NM_001339998.1 | NP_001326481.1 | 12 - 13 | 4615 | 91 | ||
NM_115839.4 | NP_191536.2 | 12 - 13 | 4586 | 91 | ||
GenBank mRNA | AY227252.1 | - | 11 - 12 | 4380 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |