Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035819.1 | NP_001030896.1 | 4 - 5 | 2207 | 110 | |
NM_001203209.1 | NP_001190138.1 | 5 - 6 | 2086 | 110 | ||
NM_001339998.1 | NP_001326481.1 | 5 - 6 | 2202 | 110 | ||
NM_115839.4 | NP_191536.2 | 5 - 6 | 2173 | 110 | ||
GenBank mRNA | AY227252.1 | - | 4 - 5 | 1967 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |