Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASHH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340008.1 | NP_001319802.1 | 6 - 7 | 746 | 100 | |
NM_001340009.1 | NP_001326226.1 | 6 - 7 | 746 | 100 | ||
NM_001340010.1 | NP_001326228.1 | 6 - 7 | 606 | 100 | ||
NM_001340011.1 | NP_001326227.1 | 6 - 7 | 606 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |