Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340024.1 | NP_001327679.1 | 3 - 4 | 481 | 103 | |
NM_115877.4 | NP_191573.1 | 3 - 4 | 481 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |