Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203212.1 | NP_001190141.1 | 5 - 6 | 594 | 72 | |
NM_001340039.1 | NP_001327362.1 | 5 - 6 | 587 | 72 | ||
NM_001340040.1 | NP_001327363.1 | 4 - 5 | 621 | 72 | ||
NM_115897.5 | NP_191592.5 | 4 - 5 | 592 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |