Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340058.1 | NP_001327227.1 | 8 - 9 | 1563 | 121 | |
NM_001340059.1 | NP_001327226.1 | 8 - 9 | 1666 | 121 | ||
NM_115938.6 | NP_191633.3 | 8 - 9 | 1823 | 121 | ||
GenBank mRNA | AF486851.1 | - | 8 - 9 | 1627 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |