Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115947.3 | NP_567105.1 | 2 - 3 | 755 | 68 | |
GenBank mRNA | AF507915.1 | - | 2 - 3 | 440 | 68 | |
AY087739.1 | - | 2 - 3 | 643 | 68 | ||
AY099661.1 | - | 2 - 3 | 665 | 68 | ||
AY128849.1 | - | 2 - 3 | 440 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |