Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PP2-A13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115970.4 | NP_567108.2 | 2 - 3 | 798 | 100 | |
NM_202741.1 | NP_974470.1 | 2 - 3 | 707 | 100 | ||
GenBank mRNA | BT000895.1 | - | 2 - 3 | 448 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |