Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RS27A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115975.6 | NP_191670.1 | 3 - 4 | 318 | 84 | |
GenBank mRNA | AF083336.1 | - | 3 - 4 | 192 | 84 | |
AF412053.1 | - | 3 - 4 | 189 | 84 | ||
AY090259.1 | - | 2 - 3 | 129 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |