Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115977.4 | NP_191672.1 | 7 - 8 | 876 | 69 | |
GenBank mRNA | DQ370437.1 | - | 6 - 7 | 474 | 69 | |
KJ138782.1 | - | 6 - 7 | 474 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |