Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115978.4 | NP_567109.1 | 2 - 3 | 872 | 110 | |
GenBank mRNA | AF360295.1 | - | 2 - 3 | 758 | 110 | |
AF395057.1 | - | 2 - 3 | 704 | 110 | ||
AY051056.1 | - | 2 - 3 | 704 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |