Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128578.4 | NP_180584.1 | 4 - 5 | 1297 | 151 | |
GenBank mRNA | AF418309.1 | - | 4 - 5 | 1048 | 151 | |
AY062720.1 | - | 4 - 5 | 1157 | 151 | ||
AY114650.1 | - | 4 - 5 | 1048 | 151 | ||
AY136318.1 | - | 4 - 5 | 1157 | 151 | ||
BT002338.1 | - | 4 - 5 | 1048 | 151 | ||
BT008482.1 | - | 4 - 5 | 1048 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |