Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2B'' | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336249.1 | NP_001324159.1 | 1 - 2 | 309 | 76 | |
NM_128579.4 | NP_180585.1 | 1 - 2 | 309 | 76 | ||
GenBank mRNA | AK227609.1 | - | 1 - 2 | 153 | 76 | |
BT003068.1 | - | 1 - 2 | 78 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |