Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121319.5 | NP_196820.1 | 1 - 2 | 472 | 77 | |
GenBank mRNA | AF314176.1 | - | 1 - 2 | 375 | 77 | |
AF372927.1 | - | 1 - 2 | 267 | 77 | ||
AY078037.1 | - | 1 - 2 | 124 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |