Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343274.1 | NP_001318555.1 | 6 - 7 | 873 | 107 | |
NM_001343275.1 | NP_001331012.1 | 6 - 7 | 804 | 107 | ||
NM_001343276.1 | NP_001331011.1 | 6 - 7 | 804 | 107 | ||
NM_121351.5 | NP_196852.3 | 5 - 6 | 840 | 107 | ||
GenBank mRNA | BT012225.1 | - | 5 - 6 | 567 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |