Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KEG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203374.1 | NP_001190303.1 | 4 - 5 | 1543 | 61 | |
NM_121356.3 | NP_196857.2 | 4 - 5 | 1867 | 61 | ||
GenBank mRNA | DQ315360.1 | - | 4 - 5 | 1451 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |