Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085109.1 | NP_001078578.1 | 2 - 3 | 1631 | 88 | |
NM_121366.4 | NP_196867.1 | 2 - 3 | 1833 | 88 | ||
GenBank mRNA | AY070133.1 | - | 2 - 3 | 1551 | 88 | |
BT002311.1 | - | 2 - 3 | 1486 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |