Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121373.4 | NP_196874.1 | 3 - 4 | 536 | 101 | |
GenBank mRNA | BT026370.1 | - | 3 - 4 | 415 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |