Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085110.1 | NP_001078579.1 | 8 - 9 | 623 | 71 | |
NM_121374.4 | NP_196875.1 | 9 - 10 | 907 | 71 | ||
GenBank mRNA | AF090372.1 | - | 9 - 10 | 574 | 71 | |
AF195648.1 | - | 9 - 10 | 585 | 71 | ||
AY120716.1 | - | 9 - 10 | 599 | 71 | ||
BT000058.1 | - | 8 - 9 | 532 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |