Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XTH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343297.1 | NP_001331376.1 | 2 - 3 | 1156 | 97 | |
NM_001343298.1 | NP_001331375.1 | 3 - 4 | 879 | 97 | ||
NM_121390.5 | NP_196891.1 | 3 - 4 | 725 | 97 | ||
GenBank mRNA | AB026486.1 | - | 2 - 3 | 323 | 97 | |
AK176228.1 | - | 2 - 3 | 303 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |