Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUVH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343303.1 | NP_001332445.1 | 2 - 3 | 575 | 97 | |
NM_121399.3 | NP_196900.1 | 2 - 3 | 534 | 97 | ||
GenBank mRNA | BT002313.1 | - | 1 - 2 | 362 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |