Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARAB-AK-LYS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085112.2 | NP_001078581.1 | 11 - 12 | 1792 | 90 | |
NM_121409.4 | NP_196910.1 | 11 - 12 | 1795 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |