Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCI11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121413.2 | NP_196914.1 | 9 - 10 | 969 | 99 | |
GenBank mRNA | AK118738.1 | - | 9 - 10 | 858 | 99 | |
AY085446.1 | - | 9 - 10 | 969 | 99 | ||
BT006054.1 | - | 9 - 10 | 758 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |