Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343319.1 | NP_001330562.1 | 2 - 3 | 447 | 88 | |
NM_001343321.1 | NP_001330560.1 | 2 - 3 | 447 | 88 | ||
NM_121422.4 | NP_568295.2 | 2 - 3 | 585 | 88 | ||
GenBank mRNA | AK176026.1 | - | 2 - 3 | 345 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Arabidopsis thaliana Probe spans exons |