Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121429.5 | NP_568296.1 | 8 - 9 | 1127 | 67 | |
GenBank mRNA | AF361760.1 | - | 8 - 9 | 1067 | 67 | |
AF395059.1 | - | 8 - 9 | 1109 | 67 | ||
AY084910.1 | - | 8 - 9 | 1105 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |