Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BET9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203379.1 | NP_001190308.1 | 2 - 3 | 1715 | 92 | |
NM_001343329.1 | NP_001332743.1 | 3 - 4 | 1496 | 92 | ||
NM_121431.3 | NP_568297.1 | 2 - 3 | 1720 | 92 | ||
GenBank mRNA | AY059785.1 | - | 2 - 3 | 1564 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |