Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121480.4 | NP_568304.1 | 2 - 3 | 620 | 93 | |
GenBank mRNA | AY045626.1 | - | 2 - 3 | 279 | 93 | |
BT001009.1 | - | 1 - 2 | 123 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |