Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121480.4 | NP_568304.1 | 6 - 7 | 1354 | 64 | |
GenBank mRNA | AY045626.1 | - | 6 - 7 | 1013 | 64 | |
BT001009.1 | - | 5 - 6 | 857 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |