Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMII | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121499.3 | NP_196999.1 | 1 - 2 | 846 | 151 | |
GenBank mRNA | DQ029214.1 | - | 1 - 2 | 722 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |