Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTSH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343392.1 | NP_001318565.1 | 3 - 4 | 1809 | 70 | |
NM_121529.2 | NP_568311.2 | 4 - 5 | 1718 | 70 | ||
GenBank mRNA | DQ446953.1 | - | 4 - 5 | 1718 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |