Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343439.1 | NP_001332392.1 | 3 - 4 | 1276 | 116 | |
NM_121623.3 | NP_197122.2 | 3 - 4 | 1276 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |