Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121662.3 | NP_568334.1 | 2 - 3 | 1483 | 99 | |
GenBank mRNA | AY030192.1 | - | 2 - 3 | 1141 | 99 | |
AY048688.1 | - | 2 - 3 | 818 | 99 | ||
BT026436.1 | - | 2 - 3 | 809 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Arabidopsis thaliana Probe spans exons |