Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLN1;4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343456.1 | NP_001331815.1 | 1 - 2 | 169 | 68 | |
NM_121663.3 | NP_568335.1 | 1 - 2 | 303 | 68 | ||
GenBank mRNA | AY059932.1 | - | 1 - 2 | 120 | 68 | |
AY128749.1 | - | 1 - 2 | 77 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |