Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036817.3 | NP_001031894.1 | 11 - 12 | 2300 | 84 | |
NM_001343458.1 | NP_001332636.1 | 10 - 11 | 2395 | 84 | ||
NM_121669.3 | NP_197166.2 | 11 - 12 | 2175 | 84 | ||
GenBank mRNA | AY062755.1 | - | 11 - 12 | 2148 | 84 | |
BT010359.1 | - | 10 - 11 | 2005 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |