Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036817.3 | NP_001031894.1 | 6 - 7 | 982 | 96 | |
NM_001343458.1 | NP_001332636.1 | 6 - 7 | 982 | 96 | ||
NM_121669.3 | NP_197166.2 | 6 - 7 | 857 | 96 | ||
GenBank mRNA | AY062755.1 | - | 6 - 7 | 830 | 96 | |
BT010359.1 | - | 5 - 6 | 687 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |