Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121681.4 | NP_568338.2 | 13 - 14 | 2400 | 94 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |