Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121681.4 | NP_568338.2 | 6 - 7 | 1376 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Arabidopsis thaliana Probe spans exons |