Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121681.4 | NP_568338.2 | 8 - 9 | 1652 | 73 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |