Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPO1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203395.1 | NP_001190324.1 | 2 - 3 | 393 | 108 | |
NM_121708.3 | NP_197204.1 | 2 - 3 | 485 | 108 | ||
GenBank mRNA | AY056126.1 | - | 2 - 3 | 381 | 108 | |
AY096633.1 | - | 2 - 3 | 213 | 108 | ||
Y18469.1 | - | 308 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |