Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPO1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203395.1 | NP_001190324.1 | 29 - 30 | 3138 | 87 | |
NM_121708.3 | NP_197204.1 | 30 - 31 | 3275 | 87 | ||
GenBank mRNA | AY056126.1 | - | 30 - 31 | 3171 | 87 | |
AY096633.1 | - | 30 - 31 | 3003 | 87 | ||
Y18469.1 | - | 3098 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |