Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121735.5 | NP_197231.1 | 6 - 7 | 745 | 102 | |
GenBank mRNA | AK117572.1 | - | 6 - 7 | 631 | 102 | |
BT005100.1 | - | 6 - 7 | 619 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |