Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121748.4 | NP_197244.1 | 10 - 11 | 2236 | 99 | |
GenBank mRNA | AY139754.1 | - | 10 - 11 | 2188 | 99 | |
BT004543.1 | - | 10 - 11 | 2143 | 99 | ||
KJ138681.1 | - | 10 - 11 | 2143 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |