Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121785.3 | NP_197281.1 | 4 - 5 | 1054 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |