Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343529.1 | NP_001331999.1 | 4 - 5 | 3082 | 66 | |
NM_121794.2 | NP_197290.1 | 4 - 5 | 2864 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |