Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121804.5 | NP_197300.1 | 1 - 2 | 467 | 86 | |
GenBank mRNA | AY054506.1 | - | 1 - 2 | 331 | 86 | |
AY093298.1 | - | 1 - 2 | 217 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |