Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125773.1 | NP_001119245.1 | 3 - 4 | 325 | 80 | |
NM_121815.3 | NP_197311.1 | 3 - 4 | 325 | 80 | ||
GenBank mRNA | AK117742.1 | - | 3 - 4 | 314 | 80 | |
BT003689.1 | - | 3 - 4 | 291 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |