Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APRL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343536.1 | NP_001330844.1 | 3 - 4 | 1215 | 103 | |
NM_001343537.1 | NP_001330845.1 | 2 - 3 | 1731 | 103 | ||
NM_121817.4 | NP_568360.1 | 3 - 4 | 941 | 103 | ||
GenBank mRNA | BT002852.1 | - | 3 - 4 | 752 | 103 | |
BT004434.1 | - | 3 - 4 | 465 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |