Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121833.7 | NP_197329.5 | 5 - 6 | 1171 | 76 | |
GenBank mRNA | AF156783.1 | - | 5 - 6 | 1126 | 76 | |
AY099866.1 | - | 5 - 6 | 1117 | 76 | ||
BT000329.1 | - | 5 - 6 | 808 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |