Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343562.1 | NP_001330131.1 | 4 - 5 | 1097 | 107 | |
NM_121873.4 | NP_197369.2 | 4 - 5 | 906 | 107 | ||
GenBank mRNA | AB493753.1 | - | 4 - 5 | 702 | 107 | |
BT010768.1 | - | 4 - 5 | 733 | 107 | ||
BT020601.1 | - | 4 - 5 | 702 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |